2-Chloro-5-[(piperidin-1-yl)methyl]pyridine 95%

Supplier: Apollo Scientific

OR32356-1G OR32356-5G
APOSOR32356-1GEA 202 GBP
APOSOR32356-1G APOSOR32356-5G
2-Chloro-5-[(piperidin-1-yl)methyl]pyridine 95%
2-Chloro-5-[(piperidin-1-yl)methyl]pyridine
Formula: C₁₁H₁₅ClN₂
MW: 210.703 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00684830
CAS Number: 230617-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR