4-(o-Tolyl)-1,2,3,6-tetrahydropyridine hydrochloride
Supplier: Apollo Scientific
OR321511-1G
OR321511-250MG
APOSOR321511-1GEA
446
GBP
APOSOR321511-1G
APOSOR321511-250MG
4-(o-Tolyl)-1,2,3,6-tetrahydropyridine hydrochloride
4-(o-Tolyl)-1,2,3,6-tetrahydropyridine hydrochloride
Formula:
C₁₂H₁₅N·ClH MW: 209,715 g/mol Storage Temperature: Refrigerator |
CAS Number:
83674-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...