4-(1H-Imidazol-1-yl)piperidine dihydrochloride 97%

Supplier: Apollo Scientific
Warning

OR321349-1G OR321349-100MG OR321349-250MG
APOSOR321349-1GEA 912 GBP
APOSOR321349-1G APOSOR321349-100MG APOSOR321349-250MG
4-(1H-Imidazol-1-yl)piperidine dihydrochloride 97%
4-(1H-Imidazol-1-yl)piperidine dihydrochloride
Formula: C₈H₁₃N₃·2ClH
MW: 224,131 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669034
CAS Number: 403492-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR