tert-Butyl (S)-2-methylpiperidine-1-carboxylate 97%

Supplier: Apollo Scientific
Warning

OR321327-1G OR321327-250MG OR321327-500MG
APOSOR321327-1GEA 1680 GBP
APOSOR321327-1G APOSOR321327-250MG APOSOR321327-500MG
tert-Butyl (S)-2-methylpiperidine-1-carboxylate 97%
tert-Butyl (S)-2-methylpiperidine-1-carboxylate
Formula: C₁₁H₂₁NO₂
MW: 199,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06799025
CAS Number: 183903-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR