4-Amino-3,5-dibromobenzoic acid 95+%
Supplier: Apollo Scientific
OR321012-10G
OR321012-5G
APOSOR321012-10GEA
178
GBP
APOSOR321012-10G
APOSOR321012-5G
4-Amino-3,5-dibromobenzoic acid 95+%
4-Amino-3,5-dibromobenzoic acid
Formula:
C₇H₅Br₂NO₂ MW: 294,928 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00075845 CAS Number: 4123-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...