1-Boc-3-acetylazetidine

Supplier: Apollo Scientific

Synonyms: tert-Butyl 3-acetylazetidine-1-carboxylate

OR317307-1G OR317307-5G
APOSOR317307-1GEA 56.3 GBP
APOSOR317307-1G APOSOR317307-5G
1-Boc-3-acetylazetidine
1-Boc-3-acetylazetidine
Formula: C₁₀H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD18073441
CAS Number: 870089-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR