5-Chloropyridine-2-carboxamide 98%

Supplier: Apollo Scientific

OR315218-1G OR315218-10G OR315218-5G
APOSOR315218-1GEA 59.1 GBP
APOSOR315218-1G APOSOR315218-10G APOSOR315218-5G
5-Chloropyridine-2-carboxamide 98%
5-Chloropyridine-2-carboxamide
Formula: C₆H₅ClN₂O
MW: 156,57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10574693
CAS Number: 370104-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR