Methyl 5-bromo-6-methoxynicotinate

Supplier: Apollo Scientific
Warning

Synonyms: 5-bromo-6-methoxy-3-pyridinecarboxylic acid methyl ester

OR313072-1G OR313072-5G
APOSOR313072-1GEA 65.6 GBP
APOSOR313072-1G APOSOR313072-5G
Methyl 5-bromo-6-methoxynicotinate
Methyl 5-bromo-6-methoxynicotinate
Formula: C₈H₈BrNO₃
MW: 246,058 g/mol
Melting Pt: 104…106 °C
Storage Temperature: Ambient
MDL Number: MFCD14698128
CAS Number: 93349-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR