3-Acetylbicyclo[1.1.1]pentane-1-carboxylic acid
Supplier: Apollo Scientific
OR312477-1G
OR312477-250MG
APOSOR312477-1GEA
2550
GBP
APOSOR312477-1G
APOSOR312477-250MG
3-Acetylbicyclo[1.1.1]pentane-1-carboxylic acid
3-Acetylbicyclo[1.1.1]pentane-1-carboxylic acid
Formula:
C₈H₁₀O₃ MW: 154,163 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18826733 CAS Number: 156329-75-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...