3-(4-Chlorophenyl)-1,4-diazaspiro[4.5]dec-3-en-2-one

Supplier: Apollo Scientific

OR311045-1G OR311045-500MG
APOSOR311045-1GEA 486 GBP
APOSOR311045-1G APOSOR311045-500MG
3-(4-Chlorophenyl)-1,4-diazaspiro[4.5]dec-3-en-2-one
3-(4-Chlorophenyl)-1,4-diazaspiro[4.5]dec-3-en-2-one
Formula: C₁₄H₁₅ClN₂O
MW: 262,735 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14735655
CAS Number: 923975-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR