Ethyl 2-amino-4-methyl-5-(morpholine-4-carbonyl)thiophene-3-carboxylate
Supplier: Apollo Scientific
OR311044-1G
OR311044-500MG
APOSOR311044-1GEA
486
GBP
APOSOR311044-1G
APOSOR311044-500MG
Ethyl 2-amino-4-methyl-5-(morpholine-4-carbonyl)thiophene-3-carboxylate
Ethyl 2-amino-4-methyl-5-(morpholine-4-carbonyl)thiophene-3-carboxylate
Formula:
C₁₃H₁₈N₂O₄S MW: 298,358 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00718001 CAS Number: 314046-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...