2,2-Dimethyl-4-phenyl-2,5-dihydro-1H-imidazole-5-thione

Supplier: Apollo Scientific

OR311043-1G OR311043-500MG
APOSOR311043-1GEA 486 GBP
APOSOR311043-1G APOSOR311043-500MG
2,2-Dimethyl-4-phenyl-2,5-dihydro-1H-imidazole-5-thione
2,2-Dimethyl-4-phenyl-2,5-dihydro-1H-imidazole-5-thione
Formula: C₁₁H₁₂N₂S
MW: 204,291 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02311186
CAS Number: 4602-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR