1-(4-Ethylphenyl)-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Supplier: Apollo Scientific

OR311034-1G OR311034-500MG
APOSOR311034-1GEA 486 GBP
APOSOR311034-1G APOSOR311034-500MG
1-(4-Ethylphenyl)-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
1-(4-Ethylphenyl)-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile
Formula: C₁₆H₁₆N₂O
MW: 252,311 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06191747
CAS Number: 727374-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR