3-[(3-Phenyl-1,2,4-oxadiazol-5-yl)methyl]-1,2,3,4-tetrahydroquinazoline-2,4-dione

Supplier: Apollo Scientific

OR311029-1G OR311029-500MG
APOSOR311029-1GEA 486 GBP
APOSOR311029-1G APOSOR311029-500MG
3-[(3-Phenyl-1,2,4-oxadiazol-5-yl)methyl]-1,2,3,4-tetrahydroquinazoline-2,4-dione
3-[(3-Phenyl-1,2,4-oxadiazol-5-yl)methyl]-1,2,3,4-tetrahydroquinazoline-2,4-dione
Formula: C₁₇H₁₂N₄O₃
MW: 320,302 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14703551
CAS Number: 892286-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR