Ethyl 1-methyl-3-(piperidine-1-sulphonyl)-1H-pyrazole-4-carboxylate

Supplier: Apollo Scientific

OR311020-1G OR311020-500MG
APOSOR311020-1GEA 486 GBP
APOSOR311020-1G APOSOR311020-500MG
Ethyl 1-methyl-3-(piperidine-1-sulphonyl)-1H-pyrazole-4-carboxylate
Ethyl 1-methyl-3-(piperidine-1-sulphonyl)-1H-pyrazole-4-carboxylate
Formula: C₁₂H₁₉N₃O₄S
MW: 301,362 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18194714
CAS Number: 1260992-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR