N-Butyl-3-(2-sulphanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide

Supplier: Apollo Scientific

OR311016-1G OR311016-500MG
APOSOR311016-1GEA 486 GBP
APOSOR311016-1G APOSOR311016-500MG
N-Butyl-3-(2-sulphanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide
N-Butyl-3-(2-sulphanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide
Formula: C₁₄H₁₇N₃OS
MW: 275,369 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14703429
CAS Number: 1146290-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR