N-(Furan-2-ylmethyl)-3-(2-sulphanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide

Supplier: Apollo Scientific

OR311015-1G OR311015-500MG
APOSOR311015-1GEA 486 GBP
APOSOR311015-1G APOSOR311015-500MG
N-(Furan-2-ylmethyl)-3-(2-sulphanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide
N-(Furan-2-ylmethyl)-3-(2-sulphanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide
Formula: C₁₅H₁₃N₃O₂S
MW: 299,348 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14703423
CAS Number: 1223880-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR