3-(4-Methoxyphenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione

Supplier: Apollo Scientific

OR311010-1G OR311010-500MG
APOSOR311010-1GEA 486 GBP
APOSOR311010-1G APOSOR311010-500MG
3-(4-Methoxyphenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione
3-(4-Methoxyphenyl)-1,4-diazaspiro[4.5]dec-3-ene-2-thione
Formula: C₁₅H₁₈N₂OS
MW: 274,381 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20528955
CAS Number: 52546-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR