[3-(Methylsulphanyl)phenyl]urea

Supplier: Apollo Scientific

OR311006-1G OR311006-500MG
APOSOR311006-1GEA 486 GBP
APOSOR311006-1G APOSOR311006-500MG
[3-(Methylsulphanyl)phenyl]urea
[3-(Methylsulphanyl)phenyl]urea
Formula: C₈H₁₀N₂OS
MW: 182,243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00175186
CAS Number: 85879-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR