1-{3-[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]pyridin-2-yl}piperidine-4-carboxylic acid

Supplier: Apollo Scientific

OR310999-1G OR310999-500MG
APOSOR310999-1GEA 486 GBP
APOSOR310999-1G APOSOR310999-500MG
1-{3-[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]pyridin-2-yl}piperidine-4-carboxylic acid
1-{3-[3-(4-Methylphenyl)-1,2,4-oxadiazol-5-yl]pyridin-2-yl}piperidine-4-carboxylic acid
Formula: C₂₀H₂₀N₄O₃
MW: 364,398 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14703505
CAS Number: 1223883-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR