2-(4-Cyclohexylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Supplier: Apollo Scientific

Synonyms: 4-Cyclohexylphenylboronic acid pinacol ester

OR310990-1G OR310990-5G
APOSOR310990-1GEA 129 GBP
APOSOR310990-1G APOSOR310990-5G
2-(4-Cyclohexylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
2-(4-Cyclohexylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₈H₂₇BO₂
MW: 286,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015819
CAS Number: 820223-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR