5-Iodo-4-methoxy-2-(propan-2-yl)phenyl 4-methylbenzene-1-sulphonate
Supplier: Apollo Scientific
OR310928-1G
OR310928-500MG
APOSOR310928-1GEA
656
GBP
APOSOR310928-1G
APOSOR310928-500MG
5-Iodo-4-methoxy-2-(propan-2-yl)phenyl 4-methylbenzene-1-sulphonate
5-Iodo-4-methoxy-2-(propan-2-yl)phenyl 4-methylbenzene-1-sulphonate
Formula:
C₁₇H₁₉IO₄S MW: 446,3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20486951 CAS Number: 865305-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...