2-(Chloromethyl)-5-methylpyridine

Supplier: Apollo Scientific
Warning

OR310896-1G OR310896-500MG
APOSOR310896-1GEA 787 GBP
APOSOR310896-1G APOSOR310896-500MG
2-(Chloromethyl)-5-methylpyridine
2-(Chloromethyl)-5-methylpyridine
Formula: C₇H₈ClN
MW: 141,598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697586
CAS Number: 767-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR