5-Iodo-2-isopropyl-4-methoxyphenol

Supplier: Apollo Scientific

OR310888-1G OR310888-500MG
APOSOR310888-1GEA 787 GBP
APOSOR310888-1G APOSOR310888-500MG
5-Iodo-2-isopropyl-4-methoxyphenol
5-Iodo-2-isopropyl-4-methoxyphenol
Formula: C₁₀H₁₃IO₂
MW: 292,113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16658587
CAS Number: 927887-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR