3-Bromo-5-chloro-1-(toluene-4-sulphonyl)-7-azaindole
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromo-5-chloro-1-[(4-methylphenyl)sulphonyl]-1H-pyrrolo[2,3-b]pyridine
OR310432-1G
OR310432-5G
OR310432-25G
APOSOR310432-1GEA
114
GBP
APOSOR310432-1G
APOSOR310432-5G
APOSOR310432-25G
3-Bromo-5-chloro-1-(toluene-4-sulphonyl)-7-azaindole
3-Bromo-5-chloro-1-(toluene-4-sulphonyl)-7-azaindole
Formula:
C₁₄H₁₀BrClN₂O₂S MW: 385,663 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11616455 CAS Number: 866546-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...