2-(tert-Butoxycarbonyl)benzylamineboronic acid

Supplier: Apollo Scientific

OR310225-1G OR310225-500MG
APOSOR310225-1GEA 656 GBP
APOSOR310225-1G APOSOR310225-500MG
2-(tert-Butoxycarbonyl)benzylamineboronic acid
2-(tert-Butoxycarbonyl)benzylamineboronic acid
Formula: C₁₂H₁₈BNO₄
MW: 251,087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663974
CAS Number: 433969-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR