4-Cyano-3,5-(ditrimethylsilyl)pyridine
Supplier: Apollo Scientific
OR310202-1G
OR310202-500MG
APOSOR310202-1GEA
551
GBP
APOSOR310202-1G
APOSOR310202-500MG
4-Cyano-3,5-(ditrimethylsilyl)pyridine
4-Cyano-3,5-(ditrimethylsilyl)pyridine
Formula:
C₁₂H₂₀N₂Si₂ MW: 248,472 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09701446 CAS Number: 827616-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...