4-(Acetoxymethyl)benzene boronic acid pinacol ester

Supplier: Apollo Scientific

OR310195-1G OR310195-500MG
APOSOR310195-1GEA 551 GBP
APOSOR310195-1G APOSOR310195-500MG
4-(Acetoxymethyl)benzene boronic acid pinacol ester
4-(Acetoxymethyl)benzene boronic acid pinacol ester
Formula: C₁₅H₂₁BO₄
MW: 276,136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179494
CAS Number: 562098-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR