2-Iodo-pivaloylaniline
Supplier: Apollo Scientific
OR310103-1G
OR310103-500MG
APOSOR310103-1GEA
328
GBP
APOSOR310103-1G
APOSOR310103-500MG
2-Iodo-pivaloylaniline
2-Iodo-pivaloylaniline
Formula:
C₁₁H₁₄INO MW: 303,139 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00859480 CAS Number: 170959-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...