N-[3-(N',N'-Dimethylamino)propyl]benzamide-3-boronic acid, pinacol ester

Supplier: Apollo Scientific

OR310097-1G OR310097-500MG
APOSOR310097-1GEA 328 GBP
APOSOR310097-1G APOSOR310097-500MG
N-[3-(N',N'-Dimethylamino)propyl]benzamide-3-boronic acid, pinacol ester
N-[3-(N',N'-Dimethylamino)propyl]benzamide-3-boronic acid, pinacol ester
Formula: C₁₈H₂₉BN₂O₃
MW: 332,245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05865085
CAS Number: 936250-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR