(2S)-4-Iodo-2-methyl-2,3-dihydropyrrolo[2,3-b]pyridine, N-BOC protected 95%

Supplier: Apollo Scientific
Warning

OR31000-1G OR31000-250MG
APOSOR31000-1GEA 72.2 GBP
APOSOR31000-1G APOSOR31000-250MG
(2S)-4-Iodo-2-methyl-2,3-dihydropyrrolo[2,3-b]pyridine, N-BOC protected 95%
(2S)-4-Iodo-2-methyl-2,3-dihydropyrrolo[2,3-b]pyridine, N-BOC protected
Formula: C₁₃H₁₇IN₂O₂
MW: 360,191 g/mol
Storage Temperature: Ambient
CAS Number: 2366997-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR