4,5,7-Trichloroquinoline
Supplier: Apollo Scientific
OR309454-1G
OR309454-250MG
APOSOR309454-1GEA
376
GBP
APOSOR309454-1G
APOSOR309454-250MG
4,5,7-Trichloroquinoline
4,5,7-Trichloroquinoline
Formula:
C₉H₄Cl₃N MW: 232,494 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07787625 CAS Number: 23834-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...