2-Amino-7-chloro-3-methylquinoline hydrochloride

Supplier: Apollo Scientific
Warning

OR309449-1G OR309449-5G
APOSOR309449-1GEA 182 GBP
APOSOR309449-1G APOSOR309449-5G
2-Amino-7-chloro-3-methylquinoline hydrochloride
2-Amino-7-chloro-3-methylquinoline hydrochloride
Formula: C₁₀H₉ClN₂·ClH
MW: 229,106 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10703334
CAS Number: 1170955-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR