4-Hydroxyquinoline-8-carboxylic acid

Supplier: Apollo Scientific
Warning

OR309341-1G OR309341-250MG
APOSOR309341-1GEA 241 GBP
APOSOR309341-1G APOSOR309341-250MG
4-Hydroxyquinoline-8-carboxylic acid
4-Hydroxyquinoline-8-carboxylic acid
Formula: C₁₀H₇NO₃
MW: 189,167 g/mol
Melting Pt: >280 °C
Storage Temperature: Ambient
MDL Number: MFCD16987425
CAS Number: 386207-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR