tert-Butyl (3R)-3-(methylamino)pyrrolidine-1-carboxylate

Supplier: Apollo Scientific

Synonyms: tert-Butyl (R)-3-(methylamino)pyrrolidine-1-carboxylate

OR308134-1G OR308134-5G
APOSOR308134-1GEA 89.2 GBP
APOSOR308134-1G APOSOR308134-5G
tert-Butyl (3R)-3-(methylamino)pyrrolidine-1-carboxylate
tert-Butyl (3R)-3-(methylamino)pyrrolidine-1-carboxylate
Formula: C₁₀H₂₀N₂O₂
MW: 200,278 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09752961
CAS Number: 199336-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR