cis-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate

Supplier: Apollo Scientific

OR307699-1G OR307699-250MG
APOSOR307699-1GEA 460 GBP
APOSOR307699-1G APOSOR307699-250MG
cis-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
cis-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
Formula: C₁₃H₁₆BrNO₂
MW: 298,176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24368283
CAS Number: 1431365-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR