trans-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
Supplier: Apollo Scientific
OR307698-1G
OR307698-250MG
APOSOR307698-1GEA
459
GBP
APOSOR307698-1G
APOSOR307698-250MG
trans-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
trans-tert-Butyl 3-(4-bromophenyl)aziridine-2-carboxylate
Formula:
C₁₃H₁₆BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD24368282 CAS Number: 1431365-04-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...