4-Cyclopropylaniline

Supplier: Apollo Scientific
Warning

OR307257-1G OR307257-5G
APOSOR307257-1GEA 110 GBP
APOSOR307257-1G APOSOR307257-5G
4-Cyclopropylaniline
4-Cyclopropylaniline
Formula: C₉H₁₁N
MW: 133.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00019219
CAS Number: 3158-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR