1-Bromo-2,5-dimethyl-4-nitrobenzene 98%
Supplier: Apollo Scientific
|
Warning
|
OR307189-1G
OR307189-250MG
APOSOR307189-1GEA
81.4
GBP
APOSOR307189-1G
APOSOR307189-250MG
1-Bromo-2,5-dimethyl-4-nitrobenzene 98%
1-Bromo-2,5-dimethyl-4-nitrobenzene
Formula:
C₈H₈BrNO₂ MW: 230,059 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01463298 CAS Number: 15540-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...