5-Bromobenzene-1,2,3-triol

Supplier: Apollo Scientific
Warning

OR305534-1G OR305534-250MG OR305534-5G
APOSOR305534-1GEA 211 GBP
APOSOR305534-1G APOSOR305534-250MG APOSOR305534-5G
5-Bromobenzene-1,2,3-triol
5-Bromobenzene-1,2,3-triol
Formula: C₆H₅BrO₃
MW: 205,006 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22555992
CAS Number: 16492-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR