4-Ethylphenylboronic acid pinacol ester

Supplier: Apollo Scientific

Synonyms: 2-(4-Ethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR303559-1G OR303559-5G
APOSOR303559-1GEA 57.7 GBP
APOSOR303559-1G APOSOR303559-5G
4-Ethylphenylboronic acid pinacol ester
4-Ethylphenylboronic acid pinacol ester
Formula: C₁₄H₂₁BO₂
MW: 232,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18643353
CAS Number: 1075719-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR