3-{4-[(4-Methylphenyl)thio]-3-nitrophenyl}acrylic acid

Supplier: Apollo Scientific

OR30332-10G OR30332-1G OR30332-5G
APOSOR30332-10GEA 570 GBP
APOSOR30332-10G APOSOR30332-1G APOSOR30332-5G
3-{4-[(4-Methylphenyl)thio]-3-nitrophenyl}acrylic acid
3-{4-[(4-Methylphenyl)thio]-3-nitrophenyl}acrylic acid
Formula: C₁₆H₁₃NO₄S
MW: 315,344 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173714
CAS Number: 175278-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR