4-Methoxy-5H,6H,7H,8H-pyrido[4,3-d]pyrimidine

Supplier: Apollo Scientific
Warning

OR303316-1G OR303316-500MG
APOSOR303316-1GEA 689 GBP
APOSOR303316-1G APOSOR303316-500MG
4-Methoxy-5H,6H,7H,8H-pyrido[4,3-d]pyrimidine
4-Methoxy-5H,6H,7H,8H-pyrido[4,3-d]pyrimidine
Formula: C₈H₁₁N₃O
MW: 165,192 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697114
CAS Number: 944902-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR