tert-Butyl 4-iodo-1H-pyrazole-1-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl 4-iodopyrazole-1-carboxylate

OR302567-1G OR302567-5G
APOSOR302567-1GEA 52.5 GBP
APOSOR302567-1G APOSOR302567-5G
tert-Butyl 4-iodo-1H-pyrazole-1-carboxylate
tert-Butyl 4-iodo-1H-pyrazole-1-carboxylate
Formula: C₈H₁₁IN₂O₂
MW: 294,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663855
CAS Number: 121669-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR