3-Chloro-2-iodoaniline 98%

Supplier: Apollo Scientific
Warning

OR301216-10G OR301216-1G OR301216-5G
APOSOR301216-10GEA 302 GBP
APOSOR301216-10G APOSOR301216-1G APOSOR301216-5G
3-Chloro-2-iodoaniline 98%
3-Chloro-2-iodoaniline
Formula: C₆H₅ClIN
MW: 253,468 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06738966
CAS Number: 70237-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR