4-Methyl-3-nitroquinoline

Supplier: Apollo Scientific
Warning

OR300364-1G OR300364-250MG
APOSOR300364-1GEA 194 GBP
APOSOR300364-1G APOSOR300364-250MG
4-Methyl-3-nitroquinoline
4-Methyl-3-nitroquinoline
Formula: C₁₀H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01574741
CAS Number: 79965-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR