2-Bromo-6,7,8,9-tetrahydro-5H-carbazole

Supplier: Apollo Scientific
Danger

OR300316-100MG OR300316-250MG
APOSOR300316-100MGEA 146 GBP
APOSOR300316-100MG APOSOR300316-250MG
2-Bromo-6,7,8,9-tetrahydro-5H-carbazole
2-Bromo-6,7,8,9-tetrahydro-5H-carbazole
Formula: C₁₂H₁₂BrN
MW: 250,134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00473712
CAS Number: 78863-99-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR