2-(1H-Pyrazol-3-yl)acetic acid

Supplier: Apollo Scientific
Warning

OR300170-1G OR300170-5G OR300170-250MG
APOSOR300170-1GEA 307 GBP
APOSOR300170-1G APOSOR300170-5G APOSOR300170-250MG
2-(1H-Pyrazol-3-yl)acetic acid
2-(1H-Pyrazol-3-yl)acetic acid
Formula: C₅H₆N₂O₂
MW: 126,113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03161180
CAS Number: 102732-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR