2-[(2-Aminoethyl)thio]ethan-1-ol 90+%
Supplier: Apollo Scientific
|
Danger
|
OR28868-10G
OR28868-1G
OR28868-5G
APOSOR28868-10GEA
492
GBP
APOSOR28868-10G
APOSOR28868-1G
APOSOR28868-5G
2-[(2-Aminoethyl)thio]ethan-1-ol 90+%
2-[(2-Aminoethyl)thio]ethan-1-ol
Formula:
C₄H₁₁NOS MW: 121,201 g/mol Boiling Pt: 97…100 °C (0,4 mm Hg) Density: 1,12 g/cm³ (25 °C) Flash Pt: 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00154135 CAS Number: 24304-84-5 EINECS: 246-142-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...