Ethyl 4-acetyl-1-cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carboxylate

Supplier: Apollo Scientific

OR25985-1G OR25985-5G
APOSOR25985-1GEA 205 GBP
APOSOR25985-1G APOSOR25985-5G
Ethyl 4-acetyl-1-cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Ethyl 4-acetyl-1-cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carboxylate
Formula: C₁₄H₁₉NO₃
MW: 249,306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407373
CAS Number: 423768-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR